Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Insulin R/CD220 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems FAB1544G
This item is not returnable.
View return policy
Description
Insulin R/CD220 Polyclonal specifically detects Insulin R/CD220 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
Insulin R/CD220 | |
Polyclonal | |
Flow Cytometry 5 uL/10^6 cells | |
CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, INSR, insulin receptor, InsulinR, IR | |
Goat | |
Affinity Purified | |
RUO | |
3643 | |
Human, Mouse | |
IgG |
Flow Cytometry | |
Alexa Fluor 488 | |
NP_001073285 | |
INSR | |
Mouse myeloma cell line NS0-derived recombinant human Insulin R/CD220 His28-Lys944 Accession # NP_001073285 | |
100 Tests | |
Primary | |
Detects human Insulin R/CD220 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse Insulin R is observed and less than 5% cross-reactivity with recombinant human INSRR is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction