Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RSK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, R&D Systems™
Rabbit Polyclonal Antibody
Supplier: R&D Systems AF1518
This item is not returnable.
View return policy
Description
RSK2 Polyclonal specifically detects RSK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RSK2 | |
Polyclonal | |
Unconjugated | |
90 kDa ribosomal protein S6 kinase 3, CLS, Coffin-Lowry syndrome, EC 2.7.11, EC 2.7.11.1, HU-3, Insulin-stimulated protein kinase 1, ISPK1, ISPK-1, MAP kinase-activated protein kinase 1b, MAPK-activated protein kinase 1b, MAPKAP kinase 1b, MAPKAPK-1b, MAPKAPK1B, mental retardation, X-linked 19, MRX19, p90-RSK2, p90RSK3, pp90RSK2, ribosomal protein S6 kinase alpha-3, ribosomal protein S6 kinase, 90kD, polypeptide 3, ribosomal protein S6 kinase, 90kDa, polypeptide 3, Ribosomal S6 kinase 2, RPS6KA3, RSK, RSK-2, RSK2p90-RSK 3, S6K-alpha3, S6K-alpha-3 | |
Rabbit | |
Affinity Purified | |
RUO | |
6197 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
RPS6KA3 | |
E. coli-derived recombinant human RSK2 | |
50 μg | |
Primary | |
Detects human, mouse, and rat RSK2 in Western blots. The antibody does not detect recombinant human RSK1 in Western blots. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction