Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT3 Goat anti-Human, Mouse, Rat, Polyclonal, R&D Systems™

Catalog No. AF1799
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1799 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1799 Supplier R&D Systems Supplier No. AF1799
This item is not returnable. View return policy

Goat Polyclonal Antibody

STAT3 Polyclonal antibody AF1799 Specifically detects STAT3 in Human, Mouse, Rat samples. It is validated for Chromatin Immunoprecipitation, Immunocytochemistry, Immunoprecipitation

Specifications

Antigen STAT3
Applications Immunoprecipitation, ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunoprecipitation 2 ug/500 ug cell lysate, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 5-15 ug/mL
Gene Accession No. U06922
Gene Alias Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor)
Gene Symbols STAT3
Host Species Goat
Immunogen E. coli-derived recombinant mouse STAT3, Arg688-Met770, Accession # U06922
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6774
Test Specificity Detects human, mouse, and rat STAT3 from whole cell extracts.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.