Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat TRRAP Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7365
This item is not returnable.
View return policy
Description
TRRAP Polyclonal specifically detects TRRAP in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
TRRAP | |
Polyclonal | |
Unconjugated | |
Q9Y4A5 | |
TRRAP | |
E. coli-derived recombinant human TRRAP Asn3610-Leu3859, predicted Accession # Q9Y4A5 | |
100 μg | |
Primary | |
Detects human, mouse and rat TRRAP in Western blots. Detects recombinant human TRRAP in direct ELISAs. | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
350/400 kDa PCAF-associated factor, FLJ10671, PAF350/400, PAF400, PAF400STAF40, STAF40, Tra1, Tra1 homolog, transformation/transcription domain-associated protein, TR-AP | |
Sheep | |
Affinity Purified | |
RUO | |
8295 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction