Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Nephrin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4269
This item is not returnable.
View return policy
Description
Nephrin Polyclonal specifically detects Nephrin in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
Nephrin | |
Polyclonal | |
Unconjugated | |
O60500 | |
NPHS1 | |
Mouse myeloma cell line NS0-derived recombinant human Nephrin Gln23-Thr1029 Accession # O60500 | |
100 μg | |
Primary | |
Detects human Nephrin in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant mouse Nephrin is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 1-15 ug/mL | |
CNF, nephrin, nephrosis 1, congenital, Finnish type (nephrin), NPHNCNF, NPHS1, Renal glomerulus-specific cell adhesion receptor | |
Sheep | |
Affinity Purified | |
RUO | |
4868 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction