Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Human Olig3, Biotin, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Supplier: R&D Systems BAF2356
This item is not returnable.
View return policy
Description
Host Species: Goat
Species Reactivity:Human
Immunogen:E. coli-derived recombinant human Olig3, Met1-Lys272, Accession No. Q7RTU3
Applications:
Immunohistochemistry, Western Blotting
Specifications
Olig3 | |
Polyclonal | |
Biotin | |
Q7RTU3 | |
OLIG3 | |
E. coli-derived recombinant human Olig3 Met1-Lys272 Accession # Q7RTU3 | |
50 μg | |
Primary | |
Detects human Olig3 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) Olig1 and rhOlig2 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Bhlhb7, BHLHE20, bHLHe20oligo3, Class B basic helix-loop-helix protein 7, Class E basic helix-loop-helix protein 20, Oligo3, oligodendrocyte transcription factor 3 | |
Goat | |
Affinity Purified | |
RUO | |
167826 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction