Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HPV Type 16 L1 Monoclonal Antibody (CAMVIR-1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA134821
Description
Recommended positive controls: HPV type 16 infected cervix. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10mM citrate buffer, pH 6.0 prior to immunostaining.
L1 protein is a major capsid protein of HPV type 16. HPV 6 and 11 have been associated with relatively benign diseases such as genital warts but type 16 and 18 are strongly associated with cervical, vaginal and vulvar malignancies.
Specifications
HPV Type 16 L1 | |
Monoclonal | |
Conc. Not Determined | |
Ascites with 0.09% sodium azide | |
Mouse | |
1 mL | |
Primary | |
4°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
CAMVIR-1 | |
Unconjugated | |
HPV L1; HPV16; HPV-16; human papillomavirus | |
BALB/C mice were immunized with recombinant HPV-16 L1 protein. | |
RUO | |
Virus | |
Antibody | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction