Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Pax7 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF6670
This item is not returnable.
View return policy
Description
Pax7 Polyclonal specifically detects Pax7 in Human samples. It is validated for Western Blot.Specifications
Pax7 | |
Polyclonal | |
Unconjugated | |
P23759 | |
PAX7 | |
E. coli-derived recombinant human Pax7 Met1-Gly196 Accession # P23759 | |
100 μg | |
Primary | |
Detects human Pax7 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) Pax3 is observed and approximately 4% cross-reactivity with rhPax1 and rhPax6 is observed. | |
Human | |
IgG |
Immunocytochemistry | |
LYOPH | |
Western Blot 0.5 μg/mL, Immunocytochemistry 5 to 25 μg/mL | |
FLJ37460, HuP1, paired box 7, paired box gene 7, paired box homeotic gene 7, paired box protein Pax-7, paired domain gene 7, PAX7 transcriptional factor, PAX7B, RMS2 | |
Goat | |
Affinity Purified | |
RUO | |
5081 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction