Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human Plexin C1, Biotin, Polyclonal, R&D Systems™

Catalog No. BAF3887
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF3887 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF3887 Supplier R&D Systems Supplier No. BAF3887

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Goat Polyclonal Antibody

Host Species: Goat
Species Reactivity:Human
Immunogen:E. coli-derived recombinant human Plexin C1 , Ile49-Ser161, Accession No. O60486

Applications:

Western Blotting

Specifications

Antigen Plexin C1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. O60486
Gene Alias CD232, CD232 antigen, plexin (semaphorin receptor), plexin C1, PLXNC1, PLXN-C1, receptor for viral semaphorin protein, receptor for virally-encoded semaphorin, VESPR, VESPRplexin-C1, Virus-encoded semaphorin protein receptor
Gene Symbols PLXNC1
Host Species Goat
Immunogen E. coli-derived recombinant human Plexin C1 Ile49-Ser161 Accession # O60486
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10154
Test Specificity Detects human Plexin C1 in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse (rm) Plexin A1, rmPlexin A2, and rmPlexin A3 is observed and less than 5% cross-reactivity with recombinant human (rh) Plexin B1 is observed, and less than 1% cross-reactivity with rhPlexin D1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.