Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plexin C1 Goat anti-Human, Polyclonal, R&D Systems™

Catalog No. AF3887
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3887 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3887 Supplier R&D Systems Supplier No. AF3887
This item is not returnable. View return policy

Goat Polyclonal Antibody

Plexin C1 Polyclonal antibody AF3887 Specifically detects Plexin C1 in Human samples. It is validated for Flow Cytometry, Western Blotting

Specifications

Antigen Plexin C1
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Gene Accession No. O60486
Gene Alias CD232, CD232 antigen, plexin (semaphorin receptor), plexin C1, PLXNC1, PLXN-C1, receptor for viral semaphorin protein, receptor for virally-encoded semaphorin, VESPR, VESPRplexin-C1, Virus-encoded semaphorin protein receptor
Gene Symbols PLXNC1
Host Species Goat
Immunogen E. coli-derived recombinant human Plexin C1 Ile49-Ser161 Accession # O60486
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10154
Test Specificity Detects human Plexin C1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant mouse (rm) Plexin A1, rmPlexin A2, rmPlexin A3, and recombinant human (rh) Plexin B1 is observed, and less than 1% cross-reactivity with rhPlexin D1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.