Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human PU.1/Spi-1, Fluorescein, Polyclonal, R&D Systems™

Catalog No. IC5870F
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC5870F 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC5870F Supplier R&D Systems Supplier No. IC5870F
This item is not returnable. View return policy

Host Species: Sheep
Species Reactivity:Human
Immunogen:E. coli-derived recombinant human PU.1, Met1-Lys169, Accession No. NP_001074016

Applications:

Flow Cytometry (Intracelloular Staining)

Specifications

Antigen PU.1/Spi-1
Applications Flow Cytometry
Classification Polyclonal
Conjugate Fluorescein
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Gene Accession No. NP_001074016
Gene Alias hematopoietic transcription factor PU.1, OF, PU.1, SFPI1, SFPI131 kDa transforming protein, SPI1, SPI-1, SPI-1 proto-oncogene, SPI-A, spleen focus forming virus (SFFV) proviral integration oncogene spi1,31 kDa-transforming protein, transcription factor PU.1
Gene Symbols SPI1
Host Species Sheep
Immunogen E. coli-derived recombinant human PU.1, Met1-Lys169, Accession # NP_001074016
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6688
Test Specificity Detects human PU.1 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human SPI-B is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.