Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFNG Sheep anti-Human, Polyclonal, R&D Systems™

Catalog No. AF7198
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7198 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7198 Supplier R&D Systems Supplier No. AF7198
This item is not returnable. View return policy

Sheep Polyclonal Antibody

RFNG Polyclonal antibody AF7198 Specifically detects RFNG in Human samples. It is validated for Western Blotting

Specifications

Antigen RFNG
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q9Y644
Gene Alias beta-1,3-N-acetylglucosaminyltransferase radical fringe, EC 2.4.1.222, O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase, radical fringe (Drosophila) homolog, radical fringe homolog, radical fringe homolog (Drosophila), RFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Gene Symbols RFNG
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human RFNG, Ala34-Arg331 (His288Gln), Accession # Q9Y644
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5986
Test Specificity Detects human RFNG in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human MFNG is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.