Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human TLR1, PE, Polyclonal, R&D Systems™

Catalog No. FAB1484P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1484P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1484P Supplier R&D Systems Supplier No. FAB1484P
This item is not returnable. View return policy

Host Species: Goat
Species Reactivity:Human
Immunogen:Mouse myeloma cell line NS0-derived recombinant human TLR1, Ser22-Asn578, Accession No. AAC34137

Applications:

Flow Cytometry

Specifications

Antigen TLR1
Applications Flow Cytometry
Classification Polyclonal
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. AAC34137
Gene Alias CD281, CD281 antigen, DKFZp547I0610, DKFZp564I0682, KIAA0012MGC126311, MGC104956, MGC126312, rsc786, TIL, TIL. LPRS5, Toll/interleukin-1 receptor-like protein, toll-like receptor 1
Gene Symbols TLR1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human TLR1 Ser22-Asn578 Accession # AAC34137
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7096
Test Specificity Detects human TLR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse (rm) TLR1 is observed and less than 5% cross-reactivity with recombinant human (rh) TLR3, rhTLR4, and rmTLR6 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.