Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human UFM1, Polyclonal, R&D Systems™

Catalog No. A500
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A500 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A500 Supplier R&D Systems Supplier No. A500
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UFM1 Polyclonal specifically detects UFM1 in Human samples. It is validated for Western Blot.

Specifications

Antigen UFM1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Gene Alias bA131P10.1, BM-002, C13orf20, chromosome 13 open reading frame 20, ubiquitin-fold modifier 1
Gene Symbols UFM1
Host Species Rabbit
Immunogen Full-length, highly purified human UFM1 protein with a 6-His tag
Purification Method Affinity Purified
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51569
Test Specificity Detects UFM1 and UFM1-protein conjugates in Western blots. Antibody does not cross-react with ubiquitin or other ubiquitin-like proteins including Nedd8, SUMO1, SUMO2, SUMO3, ISG15, FAT10 and Apg8.
Target Species Human
Content And Storage Store the unopened product at -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.