Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human XCR1, Alexa Fluor 700, Polyclonal, R&D Systems™

Catalog No. FAB857N025
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB857N025 25 Tests
FAB857N100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB857N025 Supplier R&D Systems Supplier No. FAB857N025

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Host Species: Goat
Isotype: IgG
Species Reactivity: Hu
Immunogen: E. coli-derived recombinant human XCR1. Met1-Thr31, Ser89-Lys103, His168-His190, Phe251-Tyr267

Applications:

Flow Cytometry

Specifications

Antigen XCR1/CCXCR1
Applications Flow Cytometry
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 5 uL/10^6 cells
Gene Accession No. P46094
Gene Alias CCXCR1, CCXCR1XC chemokine receptor 1, chemokine (C motif) receptor 1, chemokine XC receptor 1, G protein-coupled receptor 5, GPR5, GPR5chemokine (C motif) XC receptor 1, G-protein coupled receptor 5, Lymphotactin receptor
Gene Symbols XCR1
Host Species Goat
Immunogen E. coli-derived recombinant human XCR1, Met1-Thr31, Ser89-Lys103, His168-His190, Phe251-Tyr267, Accession # P46094
Purification Method Affinity Purified
Quantity 25 Tests
Primary or Secondary Primary
Gene ID (Entrez) 2829
Test Specificity Detects human XCR1 in direct ELISAs and Western blots.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.