Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Huntingtin Protein Mouse anti-Human, Mouse, Clone: HU-4E6, MilliporeSigma™

Catalog No. MAB2170MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2170MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2170MI Supplier MilliporeSigma Supplier No. MAB2170
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Huntingtin Protein using this Anti-Huntingtin Protein Antibody, a.a. 1247-1646, clone HU-4E6 validated for use in ELISA, immunoprecipitation, western blot, immunocytochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Huntingtin Protein
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone HU-4E6
Conjugate Unconjugated
Formulation Ascites fluid. Liquid, does not contain any preservative.
Gene Accession No. NM_002111.6
Gene Symbols HTT, IT15, SLC6A4, Huntingtin, OCD1, HD
Host Species Mouse
Immunogen Huntingtin fragment from aa 1247 to 1646 as a fusion protein
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.