Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Huntingtin Protein, Clone: mEM48, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB5374/DEL

Description
Immunocytochemistry, Immunohistochemistry, Western Blotting

Specifications
Huntingtin Protein | |
Unconjugated | |
GST fusion protein from the first 256 amino acids from human huntingtin with the deletion of the polyglutamine tract. | |
RUO | |
Primary | |
Supernatant |
Monoclonal | |
Mouse | |
100 μL | |
Neuroscience | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction