Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HuR Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07468MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-468-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-468-MI Supplier MilliporeSigma Supplier No. 07468
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-HuR Antibody is a Rabbit Polyclonal Antibody for detection of HuR also known as ELAV-like protein 1, Hu-antigen R and has been validated in immunoprecipitation, western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HuR
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation See datasheet for exact formulation.
Gene Symbols ELAVL1; MelG; HUR; ELAV1; HuR; Hua
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 1-13 of human HuR.
Purification Method See datasheet for specific purification method.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001410
Target Species Human, Mouse
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.