Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HVEM (CD270) Mouse anti-Human, Alexa Fluor 647, Clone: 94801, BD

Catalog No. BDB564411 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564411 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564411 Supplier BD Biosciences Supplier No. 564411
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 94801 monoclonal antibody specifically binds to HVEM (Herpes virus entry mediator). HVEM is also known as CD270 and LIGHT Receptor (LIGHT-R). CD270 is a type I transmembrane protein and member of the TNF Receptor superfamily. It is encoded by TNFRSF14 (tumor necrosis factor receptor superfamily member 14). CD270 is expressed on resting T cells, B cells, monocytes, and some dendritic cells. CD270 binds to homotrimeric Lymphotoxin α (LTa3/ TNFβ ), LIGHT (is homologous to lymphotoxins, exhibits inducible expression, and competes with HSV glycoprotein D for HVEM, a receptor expressed by T lymphocytes), CD272/BTLA, and glycoprotein D of Herpes simplex viruses HSV-1 and HSV-2. CD270-LIGHT interactions can reportedly transduce costimulatory signals for T cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen HVEM (CD270)
Applications Flow Cytometry
Classification Monoclonal
Clone 94801
Conjugate Alexa Fluor 647
Description LIGHT-R; HVEA; HVEM; TNFRSF14; TR2
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human HVEM Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.