Learn More
hyaluronoglucosaminidase 3, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human HYAL3 protein.
Supplier: Abnova Corporation H00008372D01P
Description
This gene encodes a protein which is similar in structure to hyaluronidases. Hyaluronidases intracellularly degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. Hyaluronan is thought to be involved in cell proliferation, migration and differentiation. However, this protein has not yet been shown to have hyaluronidase activity. The gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression. [provided by RefSeq
Sequence: MTTQLGPALVLGVALCLGCGQPLPQVPERPFSVLWNVPSAHCEARFGVHLPLNSLGIIANRGQHFHGQNMTIFYKNQLGLYPYFGPRGTAHNGGIPQALPLDRHLALAAYQIHHSLRPGFAGPAVLDWEEWCPLWAGNWGRRRAYQAASWAWAQQVFPDLDPQEQLYKAYTGFEQAARALMEDTLRVAQALRPHGLWGFYHYPACGNGWHSMASNYTGRCHAATLARNTQLHWLWAASSALFPSIYLPPRLPPAHHQAFVRHRLEEAFRVALVGHRHPLPVLAYVRLTHRRSGRFLSQDDLVQSIGVSAALGAAGVVLWGDLSLSSSEEECWHLHDYLVDTLGPYVINVTRAAMACSHQRCHGHGRCARRDPGQMEAFLHLWPDGSLGDWKSFSCHCYWGWAGPTCQEPRPGPKEAVSpecifications
hyaluronoglucosaminidase 3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human HYAL3 protein. | |
HYAL3 | |
LUCA-3/LUCA14/LUCA3/Minna14 | |
Rabbit | |
Affinity Purified | |
RUO | |
8372 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC005896.1 | |
HYAL3 | |
HYAL3 (AAH05896.1, 1 a.a. ∼ 417 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.