Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hyaluronoglucosaminidase 3, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016940 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-940 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-940 Supplier Abnova Corporation Supplier No. H00008372B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human HYAL3 protein.

This gene encodes a protein which is similar in structure to hyaluronidases. Hyaluronidases intracellularly degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. Hyaluronan is thought to be involved in cell proliferation, migration and differentiation. However, this protein has not yet been shown to have hyaluronidase activity. The gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression. [provided by RefSeq

Sequence: MTTQLGPALVLGVALCLGCGQPLPQVPERPFSVLWNVPSAHCEARFGVHLPLNSLGIIANRGQHFHGQNMTIFYKNQLGLYPYFGPRGTAHNGGIPQALPLDRHLALAAYQIHHSLRPGFAGPAVLDWEEWCPLWAGNWGRRRAYQAASWAWAQQVFPDLDPQEQLYKAYTGFEQAARALMEDTLRVAQALRPHGLWGFYHYPACGNGWHSMASNYTGRCHAATLARNTQLHWLWAASSALFPSIYLPPRLPPAHHQAFVRHRLEEAFRVALVGHRHPLPVLAYVRLTHRRSGRFLSQDDLVQSIGVSAALGAAGVVLWGDLSLSSSEEECWHLHDYLVDTLGPYVINVTRAAMACSHQRCHGHGRCARRDPGQMEAFLHLWPDGSLGDWKSFSCHCYWGWAGPTCQEPRPGPKEAV

Specifications

Antigen hyaluronoglucosaminidase 3
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human HYAL3 protein.
Formulation PBS with no preservative; pH 7.4
Gene HYAL3
Gene Accession No. BC005896.1
Gene Alias LUCA-3/LUCA14/LUCA3/Minna14
Gene Symbols HYAL3
Host Species Mouse
Immunogen HYAL3 (AAH05896.1, 1 a.a. ∼ 417 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8372
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.