Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hydroxypyruvate isomerase homolog (E. coli), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human HYI protein.
Supplier: Abnova Corporation H00081888B01
Description
Sequence: MGLGAVPGRQAAFREGLEQAVRYAKALGCPRIHLMAGRVPQGADRIAVKAEMEAVFLENLRHAAGVLAQEDLVGLLEPINTRITDPQYFLDTPQQAAAILQKVGRPNLQLQMDIFHWQIMDGNLTGNIREFLPIVGHVQVAQVPGRGEPSSPGELNFPYLFQLLEDEGYKGFVGSpecifications
hydroxypyruvate isomerase homolog (E. coli) | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human HYI protein. | |
HYI | |
HT036/MGC20767 | |
Mouse | |
50 μL | |
Yes | |
81888 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
BC019041.2 | |
HYI | |
HYI (AAH19041.1, 1 a.a. ∼ 174 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction