Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hypoxia up-regulated 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017005 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-005 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-005 Supplier Abnova Corporation Supplier No. H00010525B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human HYOU1 protein.

The protein encoded by this gene belongs to the heat shock protein 70 family. This gene uses alternative transcription start sites. A cis-acting segment found in the 5' UTR is involved in stress-dependent induction, resulting in the accumulation of this protein in the endoplasmic reticulum (ER) under hypoxic conditions. The protein encoded by this gene is thought to play an important role in protein folding and secretion in the ER. Since suppression of the protein is associated with accelerated apoptosis, it is also suggested to have an important cytoprotective role in hypoxia-induced cellular perturbation. This protein has been shown to be up-regulated in tumors, especially in breast tumors, and thus it is associated with tumor invasiveness. This gene also has an alternative translation initiation site, resulting in a protein that lacks the N-terminal signal peptide. This signal peptide-lacking protein, which is only 3 amino acids shorter than the mature protein in the ER, is thought to have a housekeeping function in the cytosol. In rat, this protein localizes to both the ER by a carboxy-terminal peptide sequence and to mitochondria by an amino-terminal targeting signal. [provided by RefSeq]

Sequence: MADKVRRQRPRRRVCWALVAVLLADLLALSDTLAVMSVDLGSESMKVAIVKPGVPMEIVLNKESRRKTPVIVTLKENERFFGDSAASMAIKNPKATLRYFQHLLGKQADNPHVALYQARFPEHELTFDPQRQTVHFQISSQLQFSPEEVLGMVLNYSRSLAEDFAEQPIKDAVITVPVFFNQAERRAVLQAARMAGLKVLQLINDNTATALSYGVFRRKDINTTAQNIMFYDMGSGSTVCTIVTYQMVKTKEAGMQPQLQIRGVGFDRTLGGLEMELRLRERLAGLFNEQRKGQRAKDVRENPRAMAKLLREANRLKTVLSANADHMAQIEGLMDDVDFKAKVTRVEFEELCADLFERVPGPVQQALQSAEMSLDEIEQVILVGGATRVPRVQEVLLKAVGKEELGKNINADEAAAMGAVYQAAALSKAFKVKPFVVRDAVVYPILVEFTREVEEEPGIHSLKHNKRVLFSRMGPYPQRKVITFNRYSHDFNFHINYGDLGFLGPEDLRVFGSQNLTTVKLKGVGDSFKKYPDYESKGIKAHFNLDESGVLSLDRVESVFETLVEDSAEEESTLTKLGNTISSLFGGGTTPDAKENGTDTVQEEEESPAEGSKDEPGEQVELKEEAEAPVEDGSQPPPPEPKGDATPEGEKATEKENGDKSEAQKPSVCRASVTRFGF

Specifications

Antigen hypoxia up-regulated 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human HYOU1 protein.
Formulation PBS with no preservative; pH 7.4
Gene HYOU1
Gene Accession No. BC072436.1
Gene Alias DKFZp686N08236/FLJ94899/FLJ97572/Grp170/HSP12A/ORP150
Gene Symbols HYOU1
Host Species Mouse
Immunogen HYOU1 (AAH72436.1, 1 a.a. ∼ 678 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10525
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.