Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HYPB Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10077334
Description
HYPB Polyclonal specifically detects HYPB in Human samples. It is validated for Flow Cytometry, Peptide ELISA.Specifications
| HYPB | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| FLJ22472, FLJ23184, FLJ45883, FLJ46217, HIF1, HIF-1EC 2.1.1.43, HIP-1, histone-lysine N-methyltransferase SETD2, hSET2, huntingtin interacting protein 1, Huntingtin yeast partner B, Huntingtin-interacting protein 1, Huntingtin-interacting protein B, HYPBp231HBP, KIAA1732SET domain-containing protein 2, KMT3AHBP231, Lysine N-methyltransferase 3A, SET domain containing 2, SET2FLJ16420 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 29072 | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, KnockDown | |
| 0.5 mg/mL | |
| Western Blot 0.5 - 2 ug/mL, Flow Cytometry 10 ug/mL, Peptide ELISA Detection limit 1:128000, Knockdown Validated | |
| NP_054878.3 | |
| SETD2 | |
| Peptide with sequence C-ERDPDKQTQNKE corresponding to internal region according to NP_054878.5. | |
| 0.1 mg | |
| Primary | |
| Expected from sequence similarity: Human, Dog, Pig. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction