Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

I-A/I-E Rat anti-Mouse, Biotin, Clone: 2G9, BD

Catalog No. BDB553622 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553622 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553622 Supplier BD Biosciences Supplier No. 553622
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 2G9 antibody reacts with the I-Ad and I-Ed MHC class II alloantigens. It also reacts with transfectants expressing I-Eb and I-Ek and with cells from mice of the H-2p and H-2q haplotypes. This antibody is non-reactive with cells from SJL (H-2s) and NOD (H-2g7) mice. Flow cytometric analysis indicates that the 2G9 and M5/114.15.2 (Cat. No. 556999, for purified mAb) monoclonal antibodies have comparable reactivity on cells from mice with I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek alloantigens.

Host Species: Rat
Clone: 2G9
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: BALB/c mouse epidermal Langerhans cells

Flow Cytometry, Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY

Specifications

Antigen I-A/I-E
Applications Flow Cytometry
Classification Monoclonal
Clone 2G9
Concentration 0.5mg/mL
Conjugate Biotin
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Rat
Immunogen BALB/c mouse epidermal Langerhans cells
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.