Learn More
I-A/I-E Rat anti-Mouse, Brilliant Violet 650, Clone: M5/114.15.2, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 563415
Description
The M5/114.15.2 monoclonal antibody recognizes a polymorphic determinant shared by the I-A[b], I-A[d], I-A[q], I-E[d], and I-E[k] (but not I-A[f], I-A[k], or I-A[s]) MHC class II alloantigens that can be expressed by B cells, dendritic cells, monocytes, macrophages and activated T cells. It also reacts with cells from mice of the H-2[p] and H-2[r] haplotypes, and it is non-reactive with cells from NOD (H-2[g7]) mice. Flow cytometric analysis indicates that the M5/114.15.2 and 2G9 monoclonal antibodies have comparable reactivity on cells from mice with I-A[b], I-A[d], I-A[g7], I-A[q], I-E[d], and I-E[k] alloantigens.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon™ BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

Specifications
I-A/I-E | |
Monoclonal | |
0.2mg/mL | |
M5/114; H-2I; Ia Ag; I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek MHC class II alloAgs | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2b κ |
Flow Cytometry | |
M5/114.15.2 | |
Brilliant Violet 650 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Activated C57BL/6 Mouse Spleen Cells | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.