Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

I-Ab Mouse anti-Mouse, FITC, Clone: AF6-120.1, BD

Catalog No. BDB562011 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562011 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562011 Supplier BD Biosciences Supplier No. 562011
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The AF6-120.1 antibody reacts with the I-A[b] MHC class II alloantigen. It cross-reacts with cells from mice of the H-2[k] and H-2[u] haplotypes. Reactivity with other haplotypes (e.g., d, f, g7, p, q, r, s) has not been observed.

TRUSTED_SUSTAINABILITY

Specifications

Antigen I-Ab
Applications Flow Cytometry
Classification Monoclonal
Clone AF6-120.1
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse C57BL/10J
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.