Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IκBα Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 071483MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-148-3MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-148-3MI Supplier MilliporeSigma Supplier No. 071483
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect IκBα using this Anti-IκBα Antibody validated for use in western blot, immunohistochemistry (paraffin), ELISA and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IκBα
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal IgG serum in buffer containing 0.02M Potassium phosphate with 0.15M NaCl, pH 7.2 and 0.1% sodium azide.
Gene Symbols NFKBIA; IKBA; MAD3; IkappaBalpha; I-kappa-B-alpha; NFKBI; IkB-alpha; MAD-3
Host Species Rabbit
Immunogen IκBα peptide corresponding to a region near the C-terminus of the human protein, conjugated to KLH.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_020529.1
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.