Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IκBα Mouse anti-Human, Unlabeled, Clone: 6A920, BD

Catalog No. BDB551819 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551819 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551819 Supplier BD Biosciences Supplier No. 551819

Mouse Monoclonal Antibody

NF-κB is a transcription factor which is a member of the mammalian NF-κB/Rel family of proteins. Members of this family are involved in the regulation of cell proliferation, immune function, as well as development. NF-κB is normally found in the cytoplasm and remains in an inactive state by its association with an inhibitory protein, IκB. Stimulation of NF-κB by a variety of inducers causes the degradation of IκBs and translocation of NF-κB to the nucleus and activation of the target gene. IκBα is a member of the IκB family of proteins, including IκBβ, IκBγ, Bcl-3, and the precursors of NF-κB1 (p105), and NF-κB2 (p100). IκBα is the best characterized member of the family and has been shown to contain three different structural domains: an N-terminal region, an amino acid internal region containing ankyrin repeats, and a C-terminal region containing a PEST domain. In resting cells, IκBα binds to and maintains NF-κB in the cytoplasm by blocking the nuclear
localization sequences of NF-κB. In response to an extracellular signal, IκBα is phosphorylated and subsequently degraded via the ubiquination-proteasome pathway, allowing NF-κB to translocate to the nucleus. Once in the nucleus, NF-κB can induce the transcription of IκBα thereby renewing the cycle so that IκBα can form a complex with NF-κB and maintain it in its cytoplasmic location. IκBα -/- mice have been shown to die soon after birth and show an increased level NF-κB activity. Furthermore, in Hodgkin's lymphoma (HL) a high constitutive level of NF-κB has been reported in samples in which clonal deleterious mutations were detected in the IκBα plays in the pathogenic process which leads to HL remains to be elucidated. IκBα migrates at ∽40kDa in SDS/PAGE, while the deduced molecular weight based upon its cDNA sequence is ∽36kDa (SWISS PROT Accession number P25963).

Host Species: Mouse
Clone: 6A920
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human IκBα aa. 105-291 tagged fusion protein

Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen IκBα
Applications Western Blot
Classification Monoclonal
Clone 6A920
Concentration 0.5mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human IκBα aa. 105-291 tagged fusion protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.