Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

I-E[k] Mouse anti-Mouse, PE, Clone: 14-4-4S, BD

Catalog No. BDB553544 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553544 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553544 Supplier BD Biosciences Supplier No. 553544
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 14-4-4S antibody reacts with the I-E[k] MHC class II alloantigen. It cross-reacts with cells from mice of the H-2[d], H-2[p], and H-2[r] haplotypes. Cells from mice of the H-2[b], H-2[f], H-2[g7], H-2[q], and H-2[s] haplotypes do not express I-E antig

Host Species: Mouse
Clone: 14-4-4S
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: C3H mouse skin graft and splenocytes

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen I-E[k]
Applications Flow Cytometry
Classification Monoclonal
Clone 14-4-4S
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen C3H mouse skin graft and splenocytes
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.