Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

I-Ek Mouse anti-Mouse, Unlabeled, Clone: 14-4-4S, BD

Catalog No. BDB558734 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558734 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558734 Supplier BD Biosciences Supplier No. 558734

Mouse Monoclonal Antibody

The 14-4-4S antibody reacts with the I-E(k) MHC class II alloantigen. It cross-reacts with cells from mice of the H-2(d), H-2(p), and H-2(r) haplotypes. Cells from mice of the H-2(b), H-2(f), H-2(g7), H-2(q), and H-2(s) haplotypes do not express I-E antigen. It has been reported that mAb 14-4-4S cross-reacts with the rat MHC class II antigen RT1D. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen I-Ek
Applications Flow Cytometry
Classification Monoclonal
Clone 14-4-4S
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen C3H mouse skin graft and splenocytes
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.