Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
I-Ek Mouse anti-Mouse, Unconjugated, Clone: 17-3-3, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 189801
Description
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
I-Ek | |
Monoclonal | |
0.5mg/mL | |
Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added | |
C3H mouse skin graft and splenocytes | |
RUO | |
Mouse | |
Liquid |
Depletion Assay, Flow Cytometry, Immunohistochemistry (Frozen) | |
17-3-3 | |
Unconjugated | |
Mouse | |
0.5 mg | |
Primary | |
Store at 2-8°C | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction