Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ia Mouse anti-Chicken, Quail, Dove, Pigeon, Duck, Biotin, Clone: CIa, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 829008
Description
SouthernBiotech is a leading provider of anti-chicken primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
| Ia | |
| Monoclonal | |
| 0.5mg/mL | |
| Supplied in PBS/NaN3 | |
| Splenic leukocytes from an outbred nine-week-old chicken | |
| RUO | |
| Chicken, Duck, Quail, Pigeon | |
| IgM |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| CIa | |
| Biotin | |
| Mouse | |
| 0.5 mg | |
| Primary | |
| Store at 2-8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction