Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IAK1 Mouse, Unlabeled, Clone: 4, BD

Catalog No. BDB610938 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610938 50 μg
BDB610939 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610938 Supplier BD Biosciences Supplier No. 610938
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cell division is a tightly regulated process that ensures the segregation of chromosomes into daughter cells. Essential to this regulation is the modification of cell cycle components by reversible phosphorylation. Ipl1 and aurora are two related kinases isolated from S.cerevisiae and Drosophila, respectively. Inactivation of these kinases results in abnormal chromosome segregation and disruption of the centrosome. A structurally and functionally similar kinase, IAK1 (Ipl1- and Aurora-related kinase 1), is a regulator of mammalian chromosome segregation. Although IAK1 may be present in the cytoplasm, it is detected on the centrosome following duplication and also associates with the spindle microtubules from metaphase through cell division. Expression of IAK1 is stringently regulated during the cell cycle. Both mRNA and protein are initially expressed in S-phase, are elevated during M-phase, and are undetectable following completion of mitosis. Increasing evidence suggests that IAK1 belongs to a novel subfamily of the ser/thr kinase superfamily. Although mutational analysis of IAK1 will directly determine its function, it appears to be a key player in the control of cell division.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen IAK1
Applications Western Blot
Classification Monoclonal
Clone 4
Concentration 250μg/mL
Conjugate Unconjugated
Description Aurora-A Kinase
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse IAK1 aa. 8-116
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.