Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IARS2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA514991
Description
This antibody is predicted to react with non-human primate based on sequence homology.
IARS2 belongs to the class-I aminoacyl-tRNA synthetase family. Members of class I have two highly conserved sequence motifs. They aminoacylate at the 2′-OH of an adenosine nucleotide, and are usually monomeric or dimeric (one or two subunits, respectively). Both classes of aminoacyl-tRNA synthetases are multidomain proteins. The catalytic domains of all the aaRSs of a given class are found to be homologous to one another, while class I and class II aaRSs are unrelated to one another. The class I aaRSs have the ubiquitous Rossmann fold and have the antiparallel beta-strands architecture while the class II aaRSs have a unique fold made up of antiparallel beta-strands.
Specifications
| IARS2 | |
| Polyclonal | |
| IARS2 | |
| Isoleucyl-tRNA synthetase, mitochondrial [Precursor]; EC 6.1.1.5; Isoleucine--tRNA ligase; IleRS; IARS2 | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 55699 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Q9NSE4 | |
| IARS2 | |
| KLH conjugated synthetic peptide between 600-628 amino acids from the central region of human IARS2 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction