Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Iba1/AIF1 Alexa Fluor 555, Clone: 20A12.1,

Catalog No. MABN92AF555 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 555
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN92AF555 100 μL Alexa Fluor 555
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN92AF555 Supplier MilliporeSigma™ Supplier No. MABN92AF555
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Iba1/AIF1 Clone: 20A12.1 in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry

Allograft inflammatory factor 1 (UniProt P55008; also known as AIF-1, Daintain, Interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, Protein G1) is encoded by the AIF1 (also known as G1, IBA1) gene (Gene ID 199) in human. Iba1/AIF1 is a 17kDa interferon-gamma- (IFN-gamma-) inducible, Ca(2+)-binding EF-hand protein. Iba1/AIF1 expression is mostly limited to the macrophage/monocyte lineage, and is augmented by cytokines, such as IFN-gamma. It is reported to be involved in neuronal function, inflammatory responses, allograft rejection, autoimmunity, and macrophage activation/function. It is also shown to be expressed in microglial cells in autominnume disesase models such as experimental autoimmune encephalomyelitis, neuritis and uveitis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Iba1/AIF1
Applications Immunohistochemistry
Classification Monoclonal
Clone 20A12.1
Conjugate Alexa Fluor 555
Formulation Purified mouse antibody conjugate in PBS with 15mg/mL BSA and 0.1 % sodium azide.
Gene Accession No. P55008
Gene Symbols AIF1, G1, IBA1
Host Species Mouse
Immunogen Linear peptide corresponding to a human Iba1/AIF1 sequence.
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001614
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.