Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anti- Ibritumomab (Anti-Idiotype) Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
| Antigen | Ibritumomab |
|---|---|
| Clone | 974622 |
| Dilution | Western Blot 2 ug/mL, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9544SP
![]() |
R&D Systems
MAB9544SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9544100
![]() |
R&D Systems
MAB9544100 |
100 μg |
Each for $468.00
|
|
|||||
Description
Ibritumomab Monoclonal specifically detects Ibritumomab in Multi-Species samples. It is validated for Western Blot, ELISA.Specifications
| Ibritumomab | |
| Western Blot 2 ug/mL, ELISA | |
| Monoclonal | |
| Supernatant | |
| Multi-species | |
| Full length Ibritumomab | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects Ibritumomab in Western blots in direct ELISAs. In direct ELISAs, 100% cross-reactivity with Tositumomab (100% sequence identity) and no cross-reactivity with Cetuximab, Rituximab, Trastuzumab, Adalimumab, Brentuximab, Gemtuzumab, Solanezumab, Teprotumumab, and Aducanumab is observed. |
| 974622 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title