Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

islet cell autoantigen 1, 69kDa, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017746 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-746 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-746 Supplier Abnova Corporation Supplier No. H00003382D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human ICA1 protein.

This gene encodes a protein with an arfaptin homology domain that is found both in the cytosol and as membrane-bound form on the Golgi complex and immature secretory granules. This protein is believed to be an autoantigen in insulin-dependent diabetes mellitus and primary Sjogren's syndrome. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq]

Sequence: MSGHKCYPWDLQDRYAQDKSVVNKMQQKYWETKQAFIKATGKKEDEHVVASDADLDAKLELFHSIQRTCLDLSKAIVLYQKRICFLSQEENELGKFLRSQGFQDKTRAGKMMQATGKALCFSSQQRLALRNPLCRFHQEVETFRHRAISDTWLTVNRMEQCRTEYRGALLWMKDVSQELDPDLYKQMEKFRKVQTQVRLAKKNFDKLKMDVCQKVDLLGASRCNLLSHMLATYQTTLLHFWEKTSHTMAAIHESFKGYQPYEFTTLKSLQDPMKKLVEKEEKKKINQQESTDAAVQEPSQLISLEEENQRKESSSFKTEDGKSILSALDKGSTHTACSGPIDELLDMKSEEGACLGPVAGTPEPEGADKDDLLLLSEIFNASSLEEGEFSKEWAAVFGDGQVKEPVPTMALGEPDPKAQTGSGFLPSQLLDQNMKDLQASLQEPAKAASDLTAWFSLFADLDPLSNPDAVGKTDKEHELLNA

Specifications

Antigen islet cell autoantigen 1, 69kDa
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human ICA1 protein.
Formulation PBS with no preservative; pH 7.4
Gene ICA1
Gene Accession No. BC008640.2
Gene Alias ICA69/ICAp69
Gene Symbols ICA1
Host Species Rabbit
Immunogen ICA1 (AAH08640.1, 1 a.a. ∼ 482 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3382
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.