Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ICAM-1/CD54, PE, Clone: 141032, R&D Systems™

Catalog No. FAB5831P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5831P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5831P Supplier R&D Systems Supplier No. FAB5831P
This item is not returnable. View return policy

Applications:

Flow Cytometry

Specifications

Antigen ICAM-1/CD54
Applications Flow Cytometry
Classification Monoclonal
Clone 141032
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q00238
Gene Alias BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM1, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
Gene Symbols ICAM1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant rat ICAM-1/CD54, Gln28-Thr493 (predicted), Accession # Q00238
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3383
Test Specificity Detects rat ICAM-1/CD54 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) DCC, recombinant human (rh) ICAM-1, rmICAM-1, rmICAM-2, rhICAM-3, rmICAM-5, rmMAdCAM-1, rhCD31 or rmVCAM-1 is observed.
Target Species Rat
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.