Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM-1 Armenian Hamster anti-Mouse, Clone: 3E2B, MilliporeSigma™

Catalog No. MAB1397ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1397ZMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1397ZMI Supplier MilliporeSigma Supplier No. MAB1397Z

Armenian Hamster Monoclonal Antibody

Anti-ICAM-1 Antibody, clone 3E2B, Azide Free detects level of ICAM-1 and has been published and validated for use in ELISA, flow cytometry, FUNC, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICAM-1
Applications ELISA, Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone 3E2B
Conjugate Unconjugated
Formulation Purified IgG prepared by affinity chromatography on Protein G agarose beads. Liquid at 1mg/mL in sterile-filtered PBS, containing no preservatives. Endotoxin level 4.13EU/mg
Gene Accession No. NM_000201.1
Gene Symbols ICAM1, P3.58, BB2, CD54, ICAM-1
Host Species Armenian Hamster
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.