Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-ICAM-1 Clone: W-CAM-1,

Catalog No. CP53100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
CP53100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CP53100UG Supplier MilliporeSigma™ Supplier No. CP53100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects ICAM-1 Clone: W-CAM-1 in Human samples, and it is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Inhibition Assays, Immuno Blotting

Cell adhesion molecules are membrane glycoproteins that play a major role in inflammation and neoplastic disease. Four primary groups of adhesion molecules have been described, which includes the integrin receptor family, the immunoglobulin superfamily, selectins, and cadherins. The intercellular adhesion molecules (ICAM-1, ICAM-2, and ICAM-3) belong to the immunoglobulin superfamily that are the counter receptors for the leukocyte integrin, lymphocyte function associated antigen 1 (LFA-1, CD11a/CD18). ICAM-1 (CD54) is a single chain glycoprotein with 7 potential N-linked glycosylation sites. It is expressed in an inducible dependent manner by endothelial and epithelial cells as well as fibroblasts, macrophages, lymphocytes, and leukocytes. ICAM-1 can mediate adhesion by binding to CD11b/CD18 (Mac-1) enhancing antigen specific T-cell activation. ICAM-1 also binds to CD43 and to Plasmodium falciparum infected erythrocytes distinct from the LFA-1 binding site. Studies indicate that ICAM-1 may also be involved in the development of atherosclerotic plaques; increased expression of ICAM-1 has been observed in all types of atherosclerotic lesions except fibrous placques. It is thought that endothelial cell expression of ICAM-1 mediates the recruitment of leukocytes to the lesion as it is suggested by its role in inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICAM-1
Applications Flow Cytometry, Immunoblot, Immunoblot, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Inhibition Assays
Classification Monoclonal
Clone W-CAM-1
Conjugate Unconjugated
Formulation In 10mM PBS, 0.2% BSA, pH 7.4.
Host Species Mouse
Immunogen Raji cells
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽85-115kDa ICAM-1 protein in endothelial cells and tonsil tissue.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.