Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

intercellular adhesion molecule 4 (Landsteiner-Wiener blood group), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016824 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-824 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-824 Supplier Abnova Corporation Supplier No. H00003386B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human ICAM4 protein.

This gene encodes the Landsteiner-Wiener (LW) blood group antigen(s) that belongs to the immunoglobulin (Ig) superfamily, and that shares similarity with the intercellular adhesion molecule (ICAM) protein family. This ICAM protein contains 2 Ig-like C2-type domains and binds to the leukocyte adhesion LFA-1 protein. The molecular basis of the LW(A)/LW(B) blood group antigens is a single aa variation at position 100; Gln-100=LW(A) and Arg-100=LW(B). Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]

Sequence: MGSLFPLSLLFFLAAAYPGVGSALGRRTKRAQSPKGSPLAPSGTSVPFWVRMSPEFVAVQPGKSVQLNCSNSCPQPQNSSLRTPLRQGKTLRGPGWVSYQLLDVRAWSSLAHCLVTCAGKTRWATSRITAYSVPGGLLGGDPEAWKPGHLFRKPGALHRPGSGQRDLDLRVCCWTPRLLAARDLPRAPQSRRPGGPQQLGTHYTDARLEPRAHSFGLRFHRCPCRDPPHCGRCVPMQVPSYEVPGVKGDVLCRLSEKKRNMKQSGEMAIHGG

Specifications

Antigen intercellular adhesion molecule 4 (Landsteiner-Wiener blood group)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human ICAM4 protein.
Formulation PBS with no preservative; pH 7.4
Gene ICAM4
Gene Accession No. NM_001039132.1
Gene Alias CD242/LW
Gene Symbols ICAM4
Host Species Mouse
Immunogen ICAM4 (NP_001034221.1, 1 a.a. ∼ 272 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3386
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.