Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICBP90 Mouse anti-Human, Unlabeled, Clone: 28, BD

Catalog No. BDB612264 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612264 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612264 Supplier BD Biosciences Supplier No. 612264
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNA topoisomerases are ubiquitous nuclear enzymes that alter the topology of DNA. Topoisomerase I produces transient single-stranded breaks in DNA, while topoisomerase II produces transient double-stranded breaks in DNA. These DNA breaks are important for DNA replication and chromosome condensation and segregation. Two types of topoisomerase II have been identified, TopoIIα and TopoIIβ. During interphase, TopoIIα is found in the nucleoplasm and nucleoli, and in mitosis, TopoIIα is found associated with metaphase chromosomes and centromeres. Transcriptional control of TopoIIα expression is regulated through multiple gene promoters, which include five inverted CCAAT boxes, one ATF binding sequence, and two GC-rich boxes. ICBP90 was identified in one-hybrid screen of proteins that can bind an inverted CCAAT box. ICBP90 is expressed highest in proliferative tissues such as thymus, liver, and bone marrow. Overexpression of ICBP90 in COS-1-transfected cells leads to enhanced expression of TopoIIα. ICBP90 co-localizes with TopoIIα in proliferating areas of appendix and inhigh grade breast carcinomas. Thus, ICBP90 may be an important transcripitonal regulator of TopoIIα expression in proliferating cells.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen ICBP90
Applications Western Blot
Classification Monoclonal
Clone 28
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human ICBP90 aa. 199-298
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.