Learn More
Invitrogen™ CD275 (B7-H2) Monoclonal Antibody (599841)

Rat Monoclonal Antibody
Supplier: Invitrogen™ MA524270
Description
In direct ELISAs, no cross-reactivity with recombinant human (rh) B7-H2, rhB7-H3, rhB7-H3b, rhPD-L2, recombinant mouse (rm) B7-1, rmB7-2, rmB7-H1, rmB7-H3, rmB7-H4, rmPD-L2, or recombinant rat B7-2 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Inducible co-stimulator ligand (ICOSL) is a specific ligand on antigen-presenting cells and cells of the peripheral tissue that binds to the inducible co-stimulator receptor (ICOS). ICOS belongs to the CD28/CD152 receptor family that regulates T-cell activation and function. ICOSL is expressed on monocytes, dendritic cells and B cells and can be induced by inflammatory stimuli in peripheral tissue. Binding to ICOSL delivers a co-stimulatory signal for T cell proliferation and cytokine secretion.
Specifications
CD275 (B7-H2) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9JHJ8 | |
Icosl | |
Mouse myeloma cell line NS0-derived recombinant mouse B7-H2 Glu47-Lys279. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry | |
599841 | |
Unconjugated | |
Icosl | |
AU044799; B7 homolog 2; B7 homologue 2; B7h; B7H2; B7-H2; B7-like protein Gl50; B7-related protein 1; B7RP1; B7RP-1; BG071784; CD275; GI50; GL50; GL50-B; ICOS ligand; ICOSL; ICOS-L; ICOSLG; inducible T-cell costimulator ligand; inducible T-cell co-stimulator ligand; KIAA0653; LICOS; Ly115l; mKIAA0653; RGD1562791; transmembrane protein B7-H2 ICOS ligand | |
Rat | |
Protein A/G | |
RUO | |
50723 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.