Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

immature colon carcinoma transcript 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017748 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-748 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-748 Supplier Abnova Corporation Supplier No. H00003396D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human ICT1 protein.

The adult colon epithelium contains 3 differentiated cell types that arise from a multipotent stem cell. Deviation from the normal maturation pathway by neoplastic transformation is thought to initiate in stem cells or their early descendants. One potential marker is ICT1 whose mRNA and protein were more highly expressed in undifferentiated than in differentiated cells. [provided by RefSeq

Sequence: MAATRCLRWGLSRAGVWLLPPPARCPRRALHKQKDGTEFKSIYSLDKLYPESQGSDTAWRVPNGAKQADSDIPLDRLTISYCRSSGPGGQNVNKVNSKAEVRFHLATAEWIAEPVRQKIAITHKNKINRLGELILTSESSRYQFRNLADCLQKIRDMITEASQTPKEPTKEDVKLHRIRIENMNRERLRQKRIHSAVKTSRRVDMD

Specifications

Antigen immature colon carcinoma transcript 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human ICT1 protein.
Formulation PBS with no preservative; pH 7.4
Gene ICT1
Gene Accession No. NM_001545.1
Gene Alias DS-1
Gene Symbols ICT1
Host Species Rabbit
Immunogen ICT1 (NP_001536.1, 1 a.a. ∼ 206 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3396
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.