Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ID-1 Mouse anti-Human, Mouse, Clone: 7D4.2, MilliporeSigma™

Catalog No. MAB4372MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4372MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4372MI Supplier MilliporeSigma Supplier No. MAB4372

Mouse Monoclonal Antibody

Detect ID-1 using this Anti-ID-1 Antibody, clone 7D4.2 validated for use in Mplex, immunofluorescence and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ID-1
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 7D4.2
Conjugate Unconjugated
Formulation Supplied in PBS Buffer, pH 7.0 with 0.1% sodium azide
Gene Accession No. NM_002165.2; NM_181353.1
Gene Symbols ID1, ID
Host Species Mouse
Immunogen Recombinant ID-1 protein
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.