Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

inhibitor of DNA binding 2, dominant negative helix-loop-helix protein, Mouse, Clone: 3C3, Abnova™

Catalog No. 89014129 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-129 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-129 Supplier Abnova Corporation Supplier No. H00003398M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant ID2.

The protein encoded by this gene belongs to the inhibitor of DNA binding (ID) family, members of which are transcriptional regulators that contain a helix-loop-helix (HLH) domain but not a basic domain. Members of the ID family inhibit the functions of basic helix-loop-helix transcription factors in a dominant-negative manner by suppressing their heterodimerization partners through the HLH domains. This protein may play a role in negatively regulating cell differentiation. A pseudogene has been identified for this gene. [provided by RefSeq

Sequence: MKAFSPVRSVRKNSLSDHSLGISRSKTPVDDPMSLLYNMNDCYSKLKELVPSIPQNKKVSKMEILQHVIDYILDLQIALDSHPTIVSLHHQRPGQNQASRTPLTTLNTDISILSLQASEFPSELMSNDSKALCG

Specifications

Antigen inhibitor of DNA binding 2, dominant negative helix-loop-helix protein
Applications ELISA, KnockDown, Western Blot
Classification Monoclonal
Clone 3C3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant ID2.
Formulation PBS with no preservative; pH 7.4
Gene ID2
Gene Accession No. BC030639
Gene Alias GIG8/ID2A/ID2H/MGC26389/bHLHb26
Gene Symbols ID2
Host Species Mouse
Immunogen ID2 (AAH30639, 1 a.a. ∼ 134 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3398
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.