Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ID3 Mouse, PE, Clone: S30-778, BD

Catalog No. BDB564564 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564564 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564564 Supplier BD Biosciences Supplier No. 564564
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The S30-778 monoclonal antibody specifically binds to Inhibitor of DNA binding 3 (ID3). ID3 is one of four ID proteins (ID1-4) that have been characterized in humans. It is also known as Class B basic helix-loop-helix protein 25 (BHLHB25), Helix-loop-helix protein HEIR-1, and ID-like protein inhibitor HLH 1R21. ID3 is a Helix-Loop-Helix (HLH) protein that can form heterodimers with other HLH transcription factors such as MyoD, myogenin, E12 and E47. These latter transcription factors normally regulate the development, differentiation and functions of a variety of cell types including muscle cells, and subsets of T and B cells. Because it lacks a basic DNA-binding domain, ID3 can inhibit the DNA binding and function of these other transcription factors it complexes with. Abnormal ID3 expression has been associated with certain cancers. The S30-778.8.66 antibody crossreacts with mouse Id3.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ID3
Applications Flow Cytometry
Classification Monoclonal
Clone S30-778
Concentration 0.2mg/mL
Conjugate PE
Description ID-3; Inhibitor of DNA binding 3; 1R21; BHLHB25; HEIR1; HEIR-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human ID3 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.