Learn More
MilliporeSigma™ anti-IDE, N-Terminal (97-273), Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ PC730100UL
Description
Specifically detects IDE, N-Terminal (97-273) Clone: in Hamster, Human, Mouse, Rat samples, and it is validated for ELISA, Immunocytochemistry, Immuno Blotting
IDE is a neutral, thiol-dependent, cytosolic, zinc metalloprotease that is reported to break downβ-amyloid.
Specifications
IDE, N-Terminal (97-273) | |
Polyclonal | |
Undiluted serum. | |
a recombinant protein consisting of amino acids 97-273 of rat IDE fused to GST | |
RUO | |
Recognizes the ∽115kDa endogenous and recombinant IDE. | |
≤ -70°C | |
IgG |
ELISA, Immunoblot, Immunoblot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
100 μL | |
Primary | |
Hamster, Human, Mouse, Rat | |
Serum |