Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

isocitrate dehydrogenase 2 (NADP+), mitochondrial, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004164 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-164 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-164 Supplier Abnova Corporation Supplier No. H00003418A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant IDH2.

Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. Each NADP(+)-dependent isozyme is a homodimer. The protein encoded by this gene is the NADP(+)-dependent isocitrate dehydrogenase found in the mitochondria. It plays a role in intermediary metabolism and energy production. This protein may tightly associate or interact with the pyruvate dehydrogenase complex. [provided by RefSeq]

Sequence: HYREHQKGRPTSTNPIASIFAWTRGLEHRGKLDGNQDLIRFAQMLEKVCVETVESGAMTKDLAGCIHGLSNVKLNEHFLNTTDFLDTIKSNLDRALGR

Specifications

Antigen isocitrate dehydrogenase 2 (NADP+), mitochondrial
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant IDH2.
Formulation 50% glycerol
Gene IDH2
Gene Accession No. NM_002168
Gene Alias ICD-M/IDH/IDHM/IDP/IDPM/mNADP-IDH
Gene Symbols IDH2
Host Species Mouse
Immunogen IDH2 (NP_002159, 354 a.a. ∼ 451 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 3418
Test Specificity This antibody cross-reacts with human IDH1.
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.